Learn More
p53 Monoclonal Antibody (PAb 240), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen AHO0112
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
This antibody reacts with only mutant p53 protein under non-denaturing conditions (Immunoprecipitation) but reacts with both mutant and wild p53 under denaturing conditions (Western blot). It reacts with an epitope at aa 212-217. Staining of formalin-fixed, paraffin-embedded tissues requires boiling tissues in 1mM EDTA, pH 8.0, for 10-20 min followed by cooling at room temperature for 20 min. Recommended positive controls include MDAD-231 cells, SK-BR-3 cells, colon carcinoma, or squamous cell carcinoma.
p53 Monoclonal antibody specifically detects p53 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
p53 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P02340, P04637 | |
Tp53, Trp53 | |
Murine p53-beta-galactosidase fusion protein containing p53 sequence from amino acid14-289. | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
PAb 240 | |
Unconjugated | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Mouse | |
Protein G | |
RUO | |
22059, 7157 | |
4° C | |
Liquid |