missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Monoclonal Antibody (X77), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA112549
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control for this product is HCT116 cell lysate.
p53 Monoclonal antibody specifically detects p53 in Human, Mouse, X. laevis samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
p53 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
P02340, P04637, P07193 | |
Tp53, tp53.L, Trp53 | |
Full-length recombinant Xenopus p53 | |
100 μg | |
Primary | |
Human, Mouse, X. laevis | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
X77 | |
Unconjugated | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Mouse | |
Protein A/G | |
RUO | |
22059, 397926, 7157 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |