missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC3 Monoclonal Antibody (J.98.6), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA514820
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
DNAJC3 Monoclonal antibody specifically detects DNAJC3 in Human, Mouse, Non-human Primate samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
DNAJC3 | |
Monoclonal | |
Unconjugated | |
DNAJC3 | |
AA408985; ACPHD; AU067833; DnaJ (Hsp40) homolog, subfamily C, member 3; DnaJ (Hsp40) homolog, subfamily C, member 3A; DnaJ (Hsp40) homolog, subfamily C, member 3B; DnaJ heat shock protein family (Hsp40) member C3; dnaJ homolog subfamily C member 3; DNAJC3; Dnajc3a; Dnajc3b; Endoplasmic reticulum DNA J domain-containing protein 6; ERdj6; ER-resident protein ERdj6; FLJ21288; HP58; Interferon-induced, double-stranded RNA-activated protein kinase inhibitor; mp58; p58; p58IPK; PRKRI; Protein kinase inhibitor of 58 kDa; protein kinase inhibitor p58; protein kinase, interferon inducible double stranded RNA dependent inhibitor; protein-kinase, interferon-inducible double stranded RNA dependent inhibitor | |
Rabbit | |
Affinity chromatography | |
RUO | |
100037258, 5611 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
J.98.6 | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
Q13217, Q91YW3 | |
DNAJC3 | |
Synthetic peptide corresponding to the sequence of human p58IPK | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |