missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NGFR Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen OSP00004W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water.
Spezifikation
NGFR | |
Polyclonal | |
NGFR | |
Low-affinity nerve growth factor receptor, NGF receptor, Gp80-LNGFR, p75 ICD, Low affinity neurotrophin receptor p75NTR, p75, LNGFR, p75NTR, RNNGFRR, CD271, p75, p-75 | |
Sheep | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P08138, P07174 | |
NGFR | |
A synthetic peptide from the extracellular domain of human p75NTR | |
RUO | |
24596, 4804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |