Learn More
PDLIM4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531151
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, HepG2. Predicted reactivity: Mouse (85%), Rat (83%), Pig (84%), Rabbit (87%), Rhesus Monkey (97%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PDLIM4 Polyclonal antibody specifically detects PDLIM4 in Human samples. It is validated for Western Blot.Spezifikation
PDLIM4 | |
Polyclonal | |
Unconjugated | |
PDLIM4 | |
enigma homolog; H-REV18; LIM domain protein; LIM protein RIL; Pdlim4; PDZ and LIM domain 4; PDZ and LIM domain protein 4; reversion induced LIM; reversion-induced LIM protein; RIL; RIT-18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P50479 | |
PDLIM4 | |
Recombinant protein fragment corresponding to a region within amino acids 2 and 228 of Human PDLIM4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |