Learn More
PEA15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543145
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: DLLTMVVDYR TRVLKISEED ELDTKLTRIP SAKKYKDIIR QPSEEEIIKL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
PEA15 Polyclonal antibody specifically detects PEA15 in Human samples. It is validated for Western Blot.Spezifikation
PEA15 | |
Polyclonal | |
Unconjugated | |
PEA15 | |
15 kDa phosphoprotein enriched in astrocytes; astrocytic phosphoprotein PEA-15; HMAT1; homolog of mouse MAT-1 oncogene; HUMMAT1H; mammary transforming gene 1; mammary transforming gene 1, mouse, homolog of; MAT1; MAT1H; OTTHUMP00000029887; Pea15; PEA-15; Pea15a; PED; phosphoprotein enriched in astrocytes 15; phosphoprotein enriched in astrocytes 15A; phosphoprotein enriched in diabetes; Pkcs15; proliferation and apoptosis adaptor protein 15 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8682 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15121 | |
PEA15 | |
Synthetic peptide directed towards the middle region of human PEA15 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |