missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Cytokeratin 18 (Ser52) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538463
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial maligcies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.Spezifikation
Phospho-Cytokeratin 18 (Ser52) | |
Polyclonal | |
Unconjugated | |
KRT18 | |
Keratin, type I cytoskeletal 18, Cell proliferation-inducing gene 46 protein, Cytokeratin-18, Short name=CK-18, Keratin-18, Short name=K18, KRT18, CYK18, PIG46, KRT 18, CK 18, CK18, Keratin 18, Keratin18, CK-18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3875 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P05783 | |
KRT18 | |
A synthetic phosphopeptide derived from human Cytokeratin 18 around the phosphorylation site of Ser52 (S-T-SP-F-R) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |