Learn More
Phospho-INSR (Tyr1158) Recombinant Polyclonal Antibody (8HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marke: Invitrogen 710101
Weitere Details : Netto-Gewicht : 0.00450kg
Beschreibung
This antibody is predicted to react with non-human primate, mouse and rat based on sequence homology. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Phospho-INSR (Tyr1158) Recombinant Polyclonal antibody specifically detects Phospho-INSR (Tyr1158) in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Phospho-INSR (Tyr1158) | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
4932439J01Rik; CD 220; CD220; CD220 antigen; CD220 beta; CD221; D630014A15Rik; HHF 5; HHF5; IGF-I receptor; INSR; Insulin receptor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; IR; IR 1; IR beta; IR1; IR-1; IR-A; IR-B | |
Phosphopeptide corresponding to amino acids 1153–1162 of human Insulin receptor | |
100 μg | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
8HCLC | |
Unconjugated | |
P06213 | |
Rabbit | |
Protein A | |
RUO | |
3643 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |