Learn More
Phospho-p53 (Ser392) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA14635
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with human and some non-human primates based on 100% sequence homology. This antibody is specific for the ∽53 kDa p53 protein phosphorylated at Ser392 in Western blots of NIH 3T3 cell lysates. Immunolabeling is blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide.
Phospho-p53 (Ser392) Polyclonal antibody specifically detects Phospho-p53 (Ser392) in Human, Rat samples. It is validated for Western Blot.Spezifikation
Phospho-p53 (Ser392) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P04637, P10361 | |
Tp53 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser392 of human TP53 conjugated to KLH | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Tp53 | |
Antigen NY-CO-13; bbl; BCC7; bfy; bhy; cellular tumor antigen p53; Cys 51 Stop; EGK_08142; FLJ92943; HGNC11998; I79_002739; LFS1; Li-Fraumeni syndrome; mutant p53; mutant tumor protein 53; OTTMUSP00000006194; p44; p53; p53 cellular tumor antigen; p53 protein; p53 tumor suppressor; p53 tumor suppressor phosphoprotein; phosphoprotein p53; Tp53; tp53.L; transformation related protein 53; transformation-related protein 53; Trp248; Trp53; tumor protein 53; tumor protein p53; tumor protein p53 (Li-Fraumeni syndrome); tumor protein p53 L homeolog; tumor suppressor p53; tumor suppressor p53 phosphoprotein; tumor suppressor protein p53; tumor supressor p53; Tumour Protein p53; XELAEV_180196761mg; Xp53; Xrel3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
24842, 7157 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |