missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PKC delta (Thr505) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517903
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with the phosphorylated forms of PKC isoforms -alpha, -beta, -gamma, -delta or -epsilon.
Phospho-PKC delta (Thr505) Polyclonal antibody specifically detects Phospho-PKC delta (Thr505) in Human, Mouse samples. It is validated for Western Blot.Spezifikation
Phospho-PKC delta (Thr505) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P28867, Q05655 | |
PRKCD | |
Synthetic phosphopeptide corresponding to residues around pThr505 of mouse PKCd | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PRKCD | |
1-May; AI385711; ALPS3; CVID9; D14Ertd420e; kinase nPKC-delta; KPCD; MAY1; nPKC-delta; PKC[d]; Pkcd; PKCdelta; Prkcd; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C[d]; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18753, 5580 | |
-20°C | |
Liquid |