missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PLD1 (Ser561) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA538390
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Phosphatidylcholine-specific phospholipases D catalyze the hydrolysis of PC to produce phosphatidic acid and choline. A range of agonists acting through G protein-coupled receptors and receptor tyrosine kinases stimulate this hydrolysis. PC-specific PLD activity has been implicated in numerous cellular pathways, including signal transduction, membrane trafficking, and the regulation of mitosis.Spezifikation
Phospho-PLD1 (Ser561) | |
Polyclonal | |
Unconjugated | |
PLD1 | |
PLD 1, choline phosphatase 1, hPLD1, phosphatidylcholine-hydrolyzing phospholipase D1, phospholipase D1, phospholipase D1, phophatidylcholine-specific | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18805, 25096, 5337 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13393, Q9Z280, P70496 | |
PLD1 | |
A synthetic phosphopeptide derived from human PLD1 around the phosphorylation site of Ser561 (K-F-SP-L-Y) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |