missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA535372
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with non-human primate based on sequence homology.
PIN1 Polyclonal antibody specifically detects PIN1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PIN1 | |
Polyclonal | |
Unconjugated | |
Pin1 | |
0610025L01Rik; D9Bwg1161e; DOD; EC 5.2.1.8; NIMA-interacting; peptidylprolyl cis/trans isomerase, NIMA-interacting 1; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 a; peptidylprolyl cis/trans isomerase, NIMA-interacting 1 S homeolog; Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1; peptidyl-prolyl cis-trans isomerase Pin1; pin1; pin1.S; pin1a; pin1-a; pin1-b; PPIase Pin1; prolyl isomerase; prolyl isomerase Pin1; protein (peptidyl-prolyl cis/trans isomerase) NIMA-interacting 1; protein (peptidylprolyl cis/trans isomerase) NIMA-interacting 1; rotamase Pin1; ubl5; XELAEV_18021616mg | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
5300 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q13526 | |
Pin1 | |
KLH conjugated synthetic peptide between 30-56 amino acids from the central region of human PIN1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |