missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINK1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518770
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with human and mouse based on sequence homology.
PINK1 Polyclonal antibody specifically detects PINK1 in Human samples. It is validated for Western Blot.Spezifikation
PINK1 | |
Polyclonal | |
Unconjugated | |
PINK1 | |
1190006F07Rik; AU042772; AW557854; BRPK; FLJ27236; mFLJ00387; PARK6; PINK 1; Pink1; PINK-1; protein kinase BRPK; PTEN induced putative kinase 1; PTEN-induced putative kinase protein 1; Serine/threonine-protein kinase PINK1, mitochondrial | |
Goat | |
Antigen affinity chromatography | |
RUO | |
65018 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9BXM7 | |
PINK1 | |
Synthetic peptide sequence (QGKAHLESRSYQEAQ) corresponding to the internal amino acids of PINK1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |