Learn More
PIP5K1C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528274
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-28274 targets PIP5K1C in WB applications and shows reactivity with Human and mouse samples. The PA5-28274 immunogen is recombit fragment corresponding to a region within amino acids 152 and 375 of Human PIP5K1C. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This gene encodes a member of the type I phosphatidylinositol-4-phosphate 5-kinase family of enzymes. A similar protein in mice is found in synapses and focal adhesion plaques, and binds the FERM domain of talin through its C-terminus.Spezifikation
PIP5K1C | |
Polyclonal | |
Unconjugated | |
O60331, O70161 | |
PIP5K1C | |
Recombit fragment corresponding to a region within amino acids 152 and 375 of Human PIP5K1C | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
0.49 mg/mL | |
PIP5K1C | |
LCCS3, PIP5K-GAMMA, PIP5Kgamma, PIPKIg_v4, Human homolog of mouse phosphatidylinositol-4-phosphate 5-kinase I-gamma, PIP5K1-gamma, diphosphoinositide kinase, phosphatidylinositol-4-phosphate 5-kinase type-1 gamma, ptdIns(4)P-5-kinase 1 gamma, type I PIP kinase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18717, 23396 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |