Learn More
SLC20A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528184
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Unboiled U87-MG, Boiled A431, Unboiled A431. Predicted reactivity: Mouse (85%), Rat (87%), Cat (90%), Rhesus Monkey (99%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the inorganic phosphate transporter family. The encoded protein is a type 3 sodium-dependent phosphate symporter that plays an important role in phosphate homeostasis by mediating cellular phosphate uptake. The encoded protein also confers susceptibility to viral infection as a gamma-retroviral receptor. Mutations in this gene may play a role in familial idiopathic basal ganglia calcification. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Spezifikation
SLC20A2 | |
Polyclonal | |
Unconjugated | |
SLC20A2 | |
Ab1-188; fi23g11; Gibbon ape leukemia virus receptor 2; Glvr2; GLVR-2; hPit2; IBGC1; IBGC3; MLVAR; MolPit2; murine leukemia virus, amphotropic, receptor for; murine leukemia virus, amphotropic; receptor; phosphate transporter 2; PIT2; PIT-2; RAM1; RAM-1; receptor for amphitropic viruses 1; receptor for amphotropic viruses 1; slc20a2; Sodium-dependent phosphate transporter 2; solute carrier family 20 (phosphate transporter), member 2; solute carrier family 20 member 2; solute carrier family 20, member 2; type III sodium-dependent phosphate transporter; wu:fi23g11; zgc:152990 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6575 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.46 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q08357 | |
SLC20A2 | |
Recombinant fragment corresponding to a region within amino acids 133 and 420 of Human PiT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |