Learn More
PIWIL4 Monoclonal Antibody (10G9B11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517151
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17151 targets PIWIL4 in indirect ELISA, FACS, IHC and WB applications and shows reactivity with Human samples. The MA5-17151 immunogen is purified recombinant fragment of human PIWIL4 (amino acids: 304-434) expressed in E. Coli. MA5-17151 detects PIWIL4 which has a predicted molecular weight of approximately 96.6kDa.
PIWIL4 Monoclonal antibody specifically detects PIWIL4 in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PIWIL4 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q7Z3Z4 | |
PIWIL4 | |
Purified recombinant fragment of human PIWIL4 (amino acids: 304-434) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
10G9B11 | |
Unconjugated | |
PIWIL4 | |
9230101H05Rik; HIWI2; mAgo5; Miwi2; PIWI; piwi like RNA-mediated gene silencing 4; Piwil4; piwi-like 4; piwi-like homolog 4; piwi-like protein 4; piwi-like RNA-mediated gene silencing 4; testis tissue sperm-binding protein Li 85P | |
Mouse | |
Protein G | |
RUO | |
143689 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |