Learn More
PKC beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529680
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: SK-N-SH, IMR32, SK-N-AS. Predicted reactivity: Mouse (98%), Rat (98%), Rabbit (98%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PKC beta Polyclonal antibody specifically detects PKC beta in Human, Mouse, Rat, X. laevis samples. It is validated for Functional Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PKC beta | |
Polyclonal | |
Unconjugated | |
PRKCB | |
A130082F03Rik; PKCB; PKC-B; PKC-beta; PRKCB; PRKCB1; PRKCB2; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; protein kinase C beta type; protein kinase C beta-II; protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18751, 25023, 5579 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Functional Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.48 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P05771, P68403, P68404 | |
PRKCB | |
Recombinant fragment corresponding to a region within amino acids 70 and 284 of Human PKC beta | |
100 μL | |
Primary | |
Human, Mouse, Rat, X. laevis | |
Antibody | |
IgG |