missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKD1L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSP00013W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water.
This gene encodes a member of the polycystin protein family containing 11 transmembrane domains, a receptor for egg jelly (REJ) domain, and a polycystin-1, lipoxygenase, alpha-toxin (PLAT) domain. The encoded protein may play a role in the male reproductive system. Alternative splice variants have been described but their biological nature has not been determined.Spezifikation
PKD1L1 | |
Polyclonal | |
PKD1L1 | |
PKD1L1, Polycystin-1L1, PK1L1 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8TDX9 | |
PKD1L1 | |
A synthetic peptide from the 1t extracellular loop of human Polycystic kidney disease protein 1-like 1 | |
RUO | |
168507 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |