Learn More
PKNOX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530244
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2 nucleus. Predicted reactivity: Mouse (94%), Rat (94%), Rhesus Monkey (99%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PKNOX1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.Spezifikation
PKNOX1 | |
Polyclonal | |
Unconjugated | |
Pknox1 | |
D17Wsu76e; homeobox protein PKNOX1; Homeobox protein PREP-1; human homeobox-containing protein; Pbx regulating protein-1; Pbx/knotted 1 homeobox; PBX/knotted 1 homeobox 1; PBX/knotted homeobox 1; PKNOX1; pkonx1c; PREP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5316 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P55347 | |
Pknox1 | |
Recombinant fragment corresponding to a region within amino acids 201 and 436 of Human PKNOX1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |