Learn More
PF4 Monoclonal Antibody (RTO), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517640
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17640 targets Platelet Factor 4 in ELISA and WB applications and shows reactivity with Human samples. The MA5-17640 immunogen is human Factor IV purified from the supernatant of thrombin activated platelets.
Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).Spezifikation
PF4 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P02776 | |
Pf4 | |
Human Factor IV purified from the supernatant of thrombin activated platelets. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
RTO | |
Biotin | |
Pf4 | |
chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4 | |
Mouse | |
Protein G | |
RUO | |
5196 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |