missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platelet Monoclonal Antibody (AIP21), FITC, Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517900
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Platelet Monoclonal antibody specifically detects Platelet in Mouse samples. It is validated for Flow Cytometry.Spezifikation
Platelet | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Rat | |
Affinity chromatography | |
RUO | |
Mouse | |
Antibody | |
IgM |
Flow Cytometry | |
AIP21 | |
FITC | |
Thrombocytes | |
Mouse Platelets | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |