Learn More
PLXNA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532026
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (96%), Zebrafish (84%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLXNA4 Polyclonal antibody specifically detects PLXNA4 in Human samples. It is validated for Western Blot.Spezifikation
PLXNA4 | |
Polyclonal | |
Unconjugated | |
Plxna4 | |
9330117B14; FAYV2820; Kiaa1550; mKIAA1550; OTTHUMP00000207245; PLEXA4; plexin A4; plexin A4, A; plexin-A4; Plxa4; Plxna4; Plxna4a; PLXNA4B; PRO34003; UNQ2820/PRO34003 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91584 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9HCM2 | |
Plxna4 | |
Recombinant fragment corresponding to a region within amino acids 19 and 261 of Human Plexin A4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |