Learn More
PLXNC1 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA547730
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse Plexin A1, A2, A3, recombinant human Plexin B1, B2, and D1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
PLXNC1 Polyclonal antibody specifically detects PLXNC1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Western Blot.Spezifikation
PLXNC1 | |
Polyclonal | |
Unconjugated | |
PLXNC1 | |
2510048K12Rik; AW742158; CD232; CD232 antigen; plexin (semaphorin receptor); plexin C1; plexin-C1; PLXNC1; PLXN-C1; receptor for viral semaphorin protein; receptor for virally-encoded semaphorin; Vespr; VESPRplexin-C1; viral-encoded semaphorin protein receptor; virus-encoded semaphorin protein receptor | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
54712 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9QZC2 | |
PLXNC1 | |
CHO-derived recombinant mouse Plexin C1 Ala35-Thr950 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |