missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLXND1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518349
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine and porcine based on sequence homology.
PLXND1 Polyclonal antibody specifically detects PLXND1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Spezifikation
PLXND1 | |
Polyclonal | |
Unconjugated | |
PLXND1 | |
6230425C21Rik; b2b1863Clo; b2b553Clo; KIAA0620; MGC75353; PLEXD1; plexin D1; plexin-D1; Plxnd1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23129 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9Y4D7 | |
PLXND1 | |
Synthetic peptide sequence (LAEPKKSHRQSH) corresponding to the internal amino acids of PLXND1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |