Learn More
PLK1 Monoclonal Antibody (3C11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517152
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-17152 targets PLK1 in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human and Mouse samples. The MA5-17152 immunogen is purified recombinant fragment of human PLK1 (amino acids: 331-508) expressed in E. Coli. MA5-17152 detects PLK1 which has a predicted molecular weight of approximately 68kDa.
PLK1 Monoclonal antibody specifically detects PLK1 in Human, Mouse samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PLK1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P53350, Q07832 | |
Plk1 | |
Purified recombinant fragment of human PLK1 (amino acids: 331-508) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
3C11 | |
Unconjugated | |
Plk1 | |
cell cycle regulated protein kinase; Plk; PLK 1; Plk1; PLK-1; plk1.S; plx1; polo (Drosophia)-like kinase; polo like kinase; polo like kinase 1; polo like kinase 1 S homeolog; polo-like kinase; polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13; STPK14; Unknown (protein for MGC:133927); XELAEV_18047692mg | |
Mouse | |
Protein G | |
RUO | |
18817, 5347 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |