Learn More
PLZF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529213
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: K562, THP-1. Predicted reactivity: Mouse (97%), Rat (96%), Chicken (85%), Rhesus Monkey (98%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PLZF Polyclonal antibody specifically detects PLZF in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
PLZF | |
Polyclonal | |
Unconjugated | |
ZBTB16 | |
AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); zinc finger protein PLZF; ZNF145 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7704 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1.03 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q05516 | |
ZBTB16 | |
Recombinant fragment corresponding to a region within amino acids 11 and 277 of Human PLZF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |