Learn More
S100A1 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA110878
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-10878 detects S100 alpha-1 (Calcium Binding Protein) from bovine and porcine. PA1-10878 has been successfully used in Western blot and immunohistochemistry applications. The PA1-10878 immunogen is Alum denatured porcine S100A1.
S100A1 Polyclonal antibody specifically detects S100A1 in Bovine, Porcine samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
S100A1 | |
Polyclonal | |
Unconjugated | |
S100A1 | |
AI266795; Protein S100-A1; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium-binding protein A1; S-100 protein alpha chain; S-100 protein subunit alpha; S100 protein, alpha polypeptide; S100a; S100A1; S100-alpha | |
Sheep | |
Ammonium sulfate precipitation | |
RUO | |
106504218, 528735 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
∽1 mg/mL | |
PBS with no preservative; pH 7.4 | |
P02639 | |
S100A1 | |
Alum denatured porcine S100A1. | |
200 μL | |
Primary | |
Bovine, Porcine | |
Antibody | |
IgG |