missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA126313
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-26313 detects S100 from human and bovine samples.
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Spezifikation
S100 | |
Polyclonal | |
Unconjugated | |
S100A1 | |
AI266795; AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-A1; protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100a; S100A1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P | |
Rabbit | |
purified | |
RUO | |
525716, 528735, 6271, 6285 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.007 mg/mL | |
PBS with 0.09% sodium azide | |
P02638, P02639, P04271, P23297 | |
S100A1, S100B | |
Affinity purified S-100 protein isolated from cow brain. | |
6 mL | |
Primary | |
Bovine, Human | |
Antibody | |
IgG |