missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100B Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA175396
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-75396 specifically detects S100B. PA1-75396 has been successfully used in ELISA applications. The PA1-75396 immunogen is Recombinant S100B.
S100B Polyclonal antibody specifically detects S100B in Human samples. It is validated for ELISA.Spezifikation
S100B | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P04271 | |
S100B | |
Recombinant human S100B | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
S100B | |
AI850290; Bpb; hypothetical protein LOC525716; NEF; protein S100-B; S100; S100 calcium binding protein B; S100 calcium binding protein beta (neural); S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S-100 calcium-binding protein beta subunit; S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S100 protein beta chain; S-100 protein beta chain; S-100 protein subunit beta; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100B; S100-B; S100beta; S100P | |
Sheep | |
Affinity chromatography | |
RUO | |
6285 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |