missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRAP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA523165
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
STRAP Polyclonal antibody specifically detects STRAP in Bovine, Canine, Equine, Human, Mouse, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
STRAP | |
Polyclonal | |
Unconjugated | |
Strap | |
AW557906; C78091; C79202; MAP activator with WD repeats; MAWD; PT-WD; serine/threonine kinase receptor associated protein; Serine-threonine kinase receptor-associated protein; STRAP; UNR-interacting protein; UNRIP; WD-40 repeat protein PT-WD | |
Rabbit | |
Protein G | |
RUO | |
100067410, 11171, 20901, 486663, 510201 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
Q5E959, Q9Y3F4, Q9Z1Z2 | |
Strap | |
A portion of amino acids 230-280 of human STRAP | |
100 μg | |
Primary | |
Bovine, Canine, Equine, Human, Mouse, Non-human Primate | |
Antibody | |
IgG |