missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUGT1 Monoclonal Antibody (1A10), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA521189
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: VVITLMIKNV QKNDVNVEFS EKELSALVKL PSGEDYNLKL ELLHPIIPEQ STFKVLSTKI EIKLKKPEAV RWEKLEGQGD VPTPKQFVAD VKNLYPSSSP YTRNWDKLVG
SUGT1 Monoclonal antibody specifically detects SUGT1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot.Spezifikation
SUGT1 | |
Monoclonal | |
0.26 mg/mL | |
PBS with no preservative; pH 7.4 | |
B0BN85, Q9CX34, Q9Y2Z0 | |
SUGT1 | |
SUGT1 (NP_006695, 151 a.a. ∽ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG3 κ |
ELISA, Immunoprecipitation, Western Blot | |
1A10 | |
Unconjugated | |
SUGT1 | |
2410174K12Rik; 40-6-3; Aa1114; LOC101122608; MGC137150 protein; Protein 40-6-3; Protein SGT1 homolog; putative 40-6-3 protein; Sgt1; SGT1 homolog, MIS12 kinetochore complex assembly cochaperone; SGT1, suppressor of G2 allele of SKP1; SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae); SGT1B protein; SUGT1; suppressor of G2 allele of SKP1; suppressor of G2 allele of SKP1 homolog; suppressor of G2 allele of SKP1, S. cerevisiae, homolog of; suppressor of g2 allele of SKP1-like protein | |
Mouse | |
Affinity chromatography | |
RUO | |
10910, 290408, 67955 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |