missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamerazine Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA175430
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-75430 specifically detects Sulfamerazine. PA1-75430 has been successfully used in ELISA applications. The PA1-75430 immunogen is Sulphamerazine-BTG.
Sulfamerazine Polyclonal antibody specifically detects Sulfamerazine in Chemical samples. It is validated for ELISA.Spezifikation
Sulfamerazine | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Sheep | |
purified | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
Unconjugated | |
C11H12N4O2S; Cremomerazine; Sulfamerazin; Sulfamethyldiazine; Sulphamerazine | |
Sulphamerazine-BTG | |
1 mg | |
Primary | |
Antibody | |
IgG |