missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT4A1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547783
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, this antibody shows less than 1% cross-reactivity with recombinant human (rh) SULT1A1, rhSULT2A1, and rhSULT1E1. Reconstitute at 0.2 mg/mL in sterile PBS.
SULT4A1 Polyclonal antibody specifically detects SULT4A1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
SULT4A1 | |
Polyclonal | |
Unconjugated | |
SULT4A1 | |
2400007A17Rik; AI853543; brain sulfotransferase 4A1; brain sulfotransferase-like protein; BRSTL1; BR-STL-1; DJ388M5.3; family 4A, member 1; hBR-STL; hBR-STL-1; LOW QUALITY PROTEIN: sulfotransferase 4A1; mBR-STL; nervous system cytosolic sulfotransferase; nervous system sulfotransferase; NST; rBR-STL; ST4A1; sulfotransferase 4A1; sulfotransferase family 4A member 1; sulfotransferase family 4A, member 1; sulfotransferase-related protein; SULT4A1; SULTX3; Unknown (protein for MGC:136828); zgc:136828 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
25830 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9BR01 | |
SULT4A1 | |
E. coli-derived recombinant human SULT4A1 isoform 1 Ala2-Leu284 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |