missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUSD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533065
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Rat (94%).
SUSD2 Polyclonal antibody specifically detects SUSD2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
SUSD2 | |
Polyclonal | |
Unconjugated | |
SUSD2 | |
BK65A6.2; SUSD2; Sushi domain (SCR repeat) containing; sushi domain containing 2; sushi domain-containing protein 2; testicular tissue protein Li 190 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56241 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9UGT4 | |
SUSD2 | |
Synthetic 17 amino acid peptide from internal region of human SUSD2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |