missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV40 T-Antigen Monoclonal Antibody (PAb416), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA190661
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody recognizes the 94 kDa SV40 large T antigen and the 20.5 kDa SV40 small T antigen. Recommended positive controls include SV80 or SV-T2 cells.
The SV40 T antigen is encoded by the early region of the SV40 genome. The large T antigen binds DNA, and complexes with a 53,000 dalton cellular protein, p53, which is required for initiation of viral DNA replication during lytic growth. In addition, the large T antigen binds DNA polymerase and the transcription factor AP-2 and forms a specific complex with the P105 product of the retinoblastoma susceptibility gene.Spezifikation
SV40 T-Antigen | |
Monoclonal | |
0.2 mg/mL | |
0.05M sodium phosphate with 0.2% gelatin and 0.1% sodium azide | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
PAb416 | |
Unconjugated | |
simian vacuolating virus 40; simian virus; Simian virus 40; SV40 T-antigen; sv40 virus large t antigen | |
Full length protein (purified) (SV40). | |
500 μL | |
Primary | |
4° C, do not freeze | |
Liquid |