Learn More
SLC22A1 Monoclonal Antibody (8C1A3), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515406
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15406 targets SYK in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15406 immunogen is purified recombinant fragment of SYK (aa296-484) expressed in E. Coli.
SLC22A1 Monoclonal antibody specifically detects SLC22A1 in Human samples. It is validated for ELISA, Western Blot.Spezifikation
SLC22A1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
O15245 | |
SLC22A1 | |
Purified recombinant fragment of SYK (aa296-484) expressed in E. Coli. | |
RUO | |
6580 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
8C1A3 | |
Unconjugated | |
SLC22A1 | |
1-Oct; DKFZp313N1010; FLJ25043; FLJ37489; H oct1_cds; hOCT1; Lx1; mOCT1; OCT1; oct1_cds; Orct; Orct1; organic cation transporter 1; rOCT1; Slc22a1; solute carrier family 22 (organic cation transporter), member 1; solute carrier family 22 member 1; solute carrier family 22, member 1; SYK | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |