missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syk Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA517812
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other members of the Syk/Zap-70 tyrosine kinase family.
Syk Polyclonal antibody specifically detects Syk in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Syk | |
Polyclonal | |
Unconjugated | |
Syk | |
DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72Syk; p72-Syk; ptk72; SP1; spleen associated tyrosine kinase; spleen tyrosine kinase; SYK; syk signaling; Sykb; tyrosine-kinase syk; tyrosine-protein kinase SYK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20963, 6850 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
73 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P43405, P48025 | |
Syk | |
Synthetic peptide corresponding to the carboxy-terminal sequence of human Syk | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |