Learn More
Synaptotagmin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544986
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: AIGKIFVGSN ATGTELRHWS DMLANPRRPI AQWHSLKPEE EVDALLGKNK Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
Synaptotagmin 2 Polyclonal antibody specifically detects Synaptotagmin 2 in Human samples. It is validated for Western Blot.Spezifikation
Synaptotagmin 2 | |
Polyclonal | |
Unconjugated | |
SYT2 | |
CMS7; Inositol polyphosphate-binding protein; IP4-binding protein; mKIAA4194; MYSPC; R74640; RATSYNII; synaptotagmin 2; synaptotagmin I; synaptotagmin II; Synaptotagmin-2; SYNII; Syt2; SytII | |
Rabbit | |
Affinity Chromatography | |
RUO | |
127833 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N9I0 | |
SYT2 | |
Synthetic peptide directed towards the C-terminal of human SYT2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |