Learn More
Syntenin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA542592
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: VGIRRAEIKQ GIREVILCKD QDGKIGLRLK SIDNGIFVQL VQANSPASLV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%
Syntenin 1 Polyclonal antibody specifically detects Syntenin 1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
Syntenin 1 | |
Polyclonal | |
Unconjugated | |
SDCBP | |
MDA9; MDA-9; melanoma differentiation associated protein-9; melanoma differentiation-associated protein 9; Pro-TGF-alpha cytoplasmic domain-interacting protein 18; sb:eu862; Scaffold protein Pbp1; sdcbp; si:dkey-235k4.1; ST1; Sycl; syndecan binding protein; syndecan binding protein (syntenin); syndecan interacting protein; syndecan-binding protein; syndecan-binding protein 1; syntenin; syntenin 1; syntenin-1; syntenin-b; TACIP18; Unknown (protein for MGC:128349) | |
Rabbit | |
Protein A | |
RUO | |
6386 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O00560 | |
SDCBP | |
Synthetic peptide directed towards the middle region of human SDCBP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |