missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase B2 Monoclonal Antibody (5026), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA143024
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-43024 detects TAFI in human samples. MA1-43024 has successfully been used in WB, ELISA, IA applications. MA1-43024 was derived from human TAFI and activated TAFI.
Carboxypeptidase B2 Monoclonal antibody specifically detects Carboxypeptidase B2 in Human samples. It is validated for ELISA, Inhibition Assays, Western Blot.Spezifikation
Carboxypeptidase B2 | |
Monoclonal | |
10.2 mg/mL | |
50% water with 50% glycerol and no preservative | |
Q96IY4 | |
CPB2 | |
Human TAFI | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Inhibition Assays, Western Blot | |
5026 | |
Unconjugated | |
CPB2 | |
1110032P04Rik; 4930405E17Rik; AI255929; carboxypeptidase B2; carboxypeptidase B2 (plasma); carboxypeptidase B2 (plasma, carboxypeptidase U); carboxypeptidase B-like protein; carboxypeptidase R; Carboxypeptidase U; Cpb2; CPR; CPU; pCPB; Plasma carboxypeptidase B; Tafi; thrombin-activable fibrinolysis inhibitor; thrombin-activatable fibrinolysis inhibitor | |
Mouse | |
Ammonium sulfate precipitation, Ion-exchange chromatography | |
RUO | |
1361 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |