missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TALDO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA513635
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TALDO1 Polyclonal antibody specifically detects TALDO1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TALDO1 | |
Polyclonal | |
Unconjugated | |
TALDO1 | |
dihydroxyacetone transferase; glycerone transferase; TAL; TALDO; TALDO1; TALDOR; TALH; TAL-H; testicular secretory protein Li 56; transaldolase; transaldolase 1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
6888 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P37837 | |
TALDO1 | |
KLH conjugated synthetic peptide between 186-215 amino acids from the central region of human TALDO1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |