missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R46 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.Spezifikation
TAS2R46 | |
Polyclonal | |
TAS2R46 | |
Taste receptor type 2 member 47, TAS2R47 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P59540 | |
TAS2R46 | |
A synthetic peptide from human taste receptor, type 2, member 47 | |
RUO | |
259292 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |