Learn More
WWTR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA14218
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-4218 detects the TAZ protein in human, mouse, and rat samples. PA1-4218 has been successfully used in Western blot procedures. By Western blot this antibody detects a ∽49 kDa protein representing TAZ in MDCK cells. The PA1-4218 immunogen is a synthetic peptide corresponding to residues V(386) E S A L N K S E P FL T W L(400) of the 49kDa human TAZ protein.
WWTR1 Polyclonal antibody specifically detects WWTR1 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
WWTR1 | |
Polyclonal | |
Unconjugated | |
WWTR1 | |
2310058J06Rik; 2610021I22Rik; C78399; DKFZp586I1419; FLJ27004; FLJ45718; Tafazzin; TAZ; transcriptional coactivator with PDZ binding motif; transcriptional co-activator with PDZ-binding motif; transcriptional coactivator with PDZ-binding motif; transcriptional co-activator with PDZ-binding motif (TA)Z; transcriptional co-activator with PDZ-binding motif (TAZ); WW domain containing transcription regulator 1; WW domain-containing transcription regulator protein 1; Wwtr1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25937, 295062, 97064 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9EPK5, Q9GZV5 | |
WWTR1 | |
Synthetic peptide corresponding to residues V(386) E S A L N K S E P FL T W L(400) of the PDZ-binding domain of human TAZ. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |