missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBPL1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518091
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.
TBPL1 Polyclonal antibody specifically detects TBPL1 in Human samples. It is validated for Western Blot.Spezifikation
TBPL1 | |
Polyclonal | |
Unconjugated | |
TBPL1 | |
21 kDa TBP-like protein; 21-kDA TBP-like protein; 4732475G08; AW011832; AW491032; D18347; MGC:8389; MGC:9620; second TBP of unique DNA protein; STD; STUD; TATA box binding protein-like 1; TATA box binding protein-like protein; TATA box-binding protein related factor 2; TATA box-binding protein-like 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1-like protein; TATA box-binding protein-related factor 2; TATA-box binding protein like 1; tbpl1; TBP-like 1; TBP-like factor; TBP-like protein; TBP-like protein 1; TBP-related factor 2; TBP-related protein; Tlf; TLP; Tlp21; trf; trf2; TRP; Unknown (protein for MGC:134257); zgc:92340 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
9519 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P62380 | |
TBPL1 | |
Synthetic peptide sequence (QIYPFVFESRKEIL) corresponding to the C-terminus amino acids of TBPL1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |