Learn More
TCP-1 delta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531523
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (85%), Xenopus laevis (90%), Chicken (94%), Chimpanzee (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TCP-1 delta Polyclonal antibody specifically detects TCP-1 delta in Human samples. It is validated for Western Blot.Spezifikation
TCP-1 delta | |
Polyclonal | |
Unconjugated | |
Cct4 | |
2610204B21Rik; A45; C78323; CCT4; cct4 protein; Cctd; CCT-delta; chaperonin containing t-complex polypeptide 1, delta subunit; chaperonin containing TCP1 subunit 4; chaperonin containing Tcp1, subunit 4 (delta); chaperonin subunit 4 (delta); SRB; Stimulator of TAR RNA-binding; T complex protein 1, delta; T-complex protein 1 subunit delta; TCP-1 delta; TCP-1-delta; Unknown (protein for MGC:134500); zgc:65789; zgc:77192 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10575 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P50991 | |
Cct4 | |
Recombinant fragment corresponding to a region within amino acids 79 and 378 of Human TCP1 delta | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |