Learn More
TCR V beta 5 Monoclonal Antibody (MEM-262), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA119020
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody recognizes beta chains of the TCR expressed by HPB-ALL cell line [carrying V(beta5.3)] and a small subset of peripheral blood T cells. This subset is larger than that recognized by other V (beta5.3)-specific antibodies. Functional Application: Activation.
TCR V beta 5 Monoclonal antibody specifically detects TCR V beta 5 in Human samples. It is validated for Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot.Spezifikation
TCR V beta 5 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
0 | |
TRB | |
Human thymoma cell line HPB-ALL. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot | |
MEM-262 | |
Unconjugated | |
TRB | |
T cell receptor beta locus; TCR V beta5; TCRB; TCRV b5; TCRV beta 5; TRB; TRB@; Vb5; Vbeta5 | |
Mouse | |
Protein A | |
RUO | |
6957 | |
4° C, do not freeze | |
Liquid |