missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR gamma/delta Monoclonal Antibody (UC7-13D5), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Marke: Invitrogen A15406
Weitere Details : Netto-Gewicht : 0.00280kg
Beschreibung
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry.Spezifikation
TCR gamma/delta | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
0 | |
Tcrd, Tcrg | |
This Armenian hamster IgG is specific to the -gamma -delta T-cell receptor (TCR) complex. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG3 |
Flow Cytometry | |
UC7-13D5 | |
FITC | |
Tcrd | |
RGD1311300; similar to T cell receptor V delta 6; T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@ | |
Armenian Hamster | |
purified | |
RUO | |
110066, 110067 | |
4° C, store in dark | |
Liquid |