missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTEX1D4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534447
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is mouse liver tissue lysate. PA5-34447 can be used with blocking peptide PEP-1489.
TCTEX1D4 Polyclonal antibody specifically detects TCTEX1D4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
TCTEX1D4 | |
Polyclonal | |
Unconjugated | |
TCTEX1D4 | |
4833401D15Rik; LOC102551208; N22.1; novel Tctex-1 family domain-containing protein; protein N22.1; Tctex1 domain containing 4; tctex1 domain-containing protein 4; tctex1 domain-containing protein 4-like; Tctex1d4; Tctex2 beta; Tctex-2-beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
102551208, 242646, 343521 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q5JR98, Q8CDY7 | |
TCTEX1D4 | |
A 19 amino acid synthetic peptide near the amino terminus of human TCTEX1D4 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |