missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA520131
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody recognizes both isoforms of TSC1. A suggested positive control is EL4 cell lysate. PA5-20131 can be used with blocking peptide PEP-0249.
TSC1 Polyclonal antibody specifically detects TSC1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
TSC1 | |
Polyclonal | |
Unconjugated | |
TSC1 | |
hamartin; HGNC:12362; Kiaa0243; LAM; MGC86987; OTTHUMP00000022439; OTTHUMP00000198378; TSC; Tsc1; tuberous sclerosis 1; tuberous sclerosis 1 protein; Tuberous sclerosis 1 protein homolog; tumor suppressor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60445, 64930, 7248 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q92574, Q9EP53, Q9Z136 | |
TSC1 | |
A 15 amino acid synthetic peptide from the middle region of human TSC1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |