missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSHZ3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534344
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is mouse brain tissue lysate. PA5-34344 can be used with blocking peptide PEP-1387.
TSHZ3 Polyclonal antibody specifically detects TSHZ3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
TSHZ3 | |
Polyclonal | |
Unconjugated | |
Tshz3 | |
A630038G13Rik; Kiaa1474; mKIAA1474; teashirt 3; teashirt homolog 3; teashirt zinc finger family member 3; teashirt zinc finger homeobox 3; teashirt3; Tsh3; Tshz3; Zfp537; zinc finger protein 537; ZNF537 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
243931, 308523, 57616 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
D3ZKB9, Q63HK5, Q8CGV9 | |
Tshz3 | |
An 18 amino acid synthetic peptide near the amino terminus of human TSHZ3 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |